Methanesulfonato[2-Bis(3,5-Di(Trifluorom 1 * 2 g
Catalog # STRE46-0340-2G
Supplier: Strem Chemicals, Inc.
Methanesulfonato[2-Bis(3,5-Di(Trifluorom 1 * 2 g
Catalog # STRE46-0340-2G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetone, BAKER, J.T.Baker®
Supplier: Avantor
N,N-Dimethylacetamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.7%, pure
Supplier: Thermo Fisher Scientific
Sinapic acid, Supelco®
Supplier: Merck
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
USP SOLUTION METHYL ORANGE 1 * 100 mL
Supplier: REAGECON
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
2-Propanol 70 % pure 1 * 1 L
Supplier: PanReac AppliChem