Ruthenium(Iii) Chloride, Anhydrous 1 * 5 g
Catalog # STRE44-5850-5G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Potassium hydrogen phthalate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium salicylate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Copper(II) sulphate pentahydrate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Sodium hydrogen carbonate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Copper(II) sulphate, anhydrous ≥99.0%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ammonium chloride 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Potassium acetate 99.0-101.0% Ph. Eur.
Supplier: VWR Chemicals