Chloro(Cyclopentadienyl){1,3-Bis[2-(Diph 1 * 100 mg
Catalog # STRE28-1095-100MG
Supplier: Strem Chemicals, Inc.
Chloro(Cyclopentadienyl){1,3-Bis[2-(Diph 1 * 100 mg
Catalog # STRE28-1095-100MG
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
1-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Indole for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone ≥99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine 99.5+% for spectroscopy
Supplier: Thermo Fisher Scientific
2-Ethoxyethanol 99%, Reagent Grade
Supplier: Honeywell Chemicals
Chloroform stabilized with ethanol pure 1 * 1.000 mL
Supplier: PanReac AppliChem
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific