Callery Potassium Hexamethyldisilazane, 1 * 100 g
Catalog # STRE19-1045-100G
Supplier: Strem Chemicals, Inc.
Callery Potassium Hexamethyldisilazane, 1 * 100 g
Catalog # STRE19-1045-100G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
tert-Butyl methyl ether, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
N,N-Dimethylacetamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Bromoform 99+% stabilised
Supplier: Thermo Fisher Scientific
Ferrozine mono-sodium salt hydrate, Supelco®
Supplier: Merck
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), Supelco®
Supplier: Merck
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific