Tris(3,5-Dimethylphenyl)Phosphine, 98% 1 * 2 g
Catalog # STRE15-7820-2G
Supplier: Strem Chemicals, Inc.
Tris(3,5-Dimethylphenyl)Phosphine, 98% 1 * 2 g
Catalog # STRE15-7820-2G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals