4C TEMPSHELL-ELEMENT–PAAR 1 * 1,1 kg
Catalog # DLTTA100029
Supplier: DELTA T
Frequently Bought Together
Disodium hydrogen phosphate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Copper(II) sulphate, anhydrous ≥99.0%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Ammonium peroxodisulphate (APS) ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium dihydrogen phosphate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium acetate 99.0-101.0% Ph. Eur.
Supplier: VWR Chemicals
Sodium dihydrogen phosphate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium sulphite, anhydrous 98-100.0% (by iodometry), Puriss. p.a. Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals