1-(2,4,6-Trimethylphenyl)-2-(Dicyclohexy 1 * 2 g
Catalog # STRE15-6362-2G
Supplier: Strem Chemicals, Inc.
1-(2,4,6-Trimethylphenyl)-2-(Dicyclohexy 1 * 2 g
Catalog # STRE15-6362-2G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for UHPLC-MS
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Dimethyl pyrocarbonate for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol : Water (30:70) % (v/v), LiChrosolv®, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
BACKGROUND BUSTER BIOSCIENDES 1 * 50 mL
Supplier: MARKET SOURCE PART PROCESS
Dichloromethane 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
Ethanol absolute, anhydrous ≥99.8% (by GC), AR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetone ≥99.5%, Environmental Grade
Supplier: Thermo Fisher Scientific