Bis(2-Cyanoethyl)Phenylphosphine, Min. 9 1 * 5 g
Catalog # STRE15-0075-5G
Supplier: Strem Chemicals, Inc.
Bis(2-Cyanoethyl)Phenylphosphine, Min. 9 1 * 5 g
Catalog # STRE15-0075-5G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Acetonitrile ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Cyclohexane ≥99.5%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Dichloromethane, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
1-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals