(S)-(-)-2,2'-Bis[Di(3,5-Di-T-Butylphenyl 1 * 2 g
Catalog # STRE15-0045-2G
Supplier: Strem Chemicals, Inc.
(S)-(-)-2,2'-Bis[Di(3,5-Di-T-Butylphenyl 1 * 2 g
Catalog # STRE15-0045-2G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Xylenes (isomeric mixture) EMPLURA® 1 * 2,5 L
Supplier: Merck
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone ≥99%
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific