(2S)-N-[(1S,2R)-2-Hydroxy-1,2-Diphenylet 1 * 100 mg
Catalog # STRE07-0983-100MG
Supplier: Strem Chemicals, Inc.
(2S)-N-[(1S,2R)-2-Hydroxy-1,2-Diphenylet 1 * 100 mg
Catalog # STRE07-0983-100MG
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitrile with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethylene glycol, high purity
Supplier: VWR Chemicals
Methanol, J.T.Baker®
Supplier: Avantor
Ethyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water for analysis
Supplier: Thermo Fisher Scientific
Isooctane for UV, IR, HPLC, ACS 1 * 1 L
Supplier: PanReac AppliChem
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific
Isobutanol, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol 99+%, extra pure
Supplier: Thermo Fisher Scientific
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific