Carbon Nanotubes, Multi-Walled, Ground C 1 * 250 mg
STRE06-0506-250MG
Frequently Bought Together
Sodium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ammonium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Citric acid, anhydrous ≥99.5%, powder, GPR RECTAPUR®
: VWR Chemicals
Ammonium metavanadate ≥99.0% analytical reagent
: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals