Fullerenes - C60/C70 Mixture (Contains ~ 1 * 50 mg
Catalog # STRE06-0500-50MG
Supplier: Strem Chemicals, Inc.
Fullerenes - C60/C70 Mixture (Contains ~ 1 * 50 mg
Catalog # STRE06-0500-50MG
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Diethyl ether ≥99% stabilised, GPR RECTAPUR® peroxide-free
Supplier: VWR Chemicals
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Supplier: Merck
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
n-Pentane, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Butyl acetate ≥99%
Supplier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
4-Methyl-2-pentanone ≥97%, TECHNICAL
Supplier: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Diethyl ether
Supplier: Roth Carl
Cyclohexane, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Acetone, J.T.Baker®
Supplier: Avantor
Petroleum spirit, 40…60 °C for residue analysis, suitable for PAH analysis
Supplier: Thermo Fisher Scientific