(1S)-2,2'-Dibromo-1,1'-Binaphthalene, Mi 1 * 100 mg
Catalog # STRE06-0484-100MG
Supplier: Strem Chemicals, Inc.
(1S)-2,2'-Dibromo-1,1'-Binaphthalene, Mi 1 * 100 mg
Catalog # STRE06-0484-100MG
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
N-Methyl-2-pyrrolidone (NMP) 98%, pure
Supplier: Thermo Fisher Scientific
Cyclohexane, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific