Orders LinkContactUsLinkComponent
Hanging racks
Hanging racks
Hanging rack, 15 hooks, 980W×160H mm
Catalog # TEKMWR0315
Supplier:   TEKNOMEK
Hanging racks
Catalog # TEKMWR0315
Supplier:  TEKNOMEK
Hanging racks
Hanging racks
Hanging rack, 15 hooks, 980W×160H mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Our stainless-steel hanging racks are manufactured to the highest standards.
  • Brushed dull polish finish
  • Wall mounted
  • Optional number etching if required
Our hanging racks are manufactured with 304 grade stainless steel for optimised durability and strength.
Long-lasting and sturdy, they are designed to the highest standard, ensuring super-smooth edges to minimise scratching and snagging.
Wall mounted, enabling space-efficiency. As with all our products, our stainless-steel coat racks are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durable and easy to clean product ideal for your workspace.
Available in a variety of sizes.
Specifications
  • Description:
    Hanging rack, 15 hooks, 980W×160H mm
  • Seq:
    07