Hanging racks
Hanging rack, 12 hooks, 1500W×95H mm
Catalog # TEKMCRGS-HR12H
Supplier: TEKNOMEK
Jump to:
Product Details & Documents
Our stainless-steel hanging racks are manufactured to the highest standards.
- Brushed dull polish finish
- Wall mounted
- Optional number etching if required
Our hanging racks are manufactured with 304 grade stainless steel for optimised durability and strength.
Long-lasting and sturdy, they are designed to the highest standard, ensuring super-smooth edges to minimise scratching and snagging.
Wall mounted, enabling space-efficiency. As with all our products, our stainless-steel coat racks are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durable and easy to clean product ideal for your workspace.
Available in a variety of sizes.
Specifications
- Description:Hanging rack, 12 hooks, 1500W×95H mm
- Seq:06