Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Mutiple-Use Pump Chamber; 316L machined stainless steel multiuse Quattroflow pump systems
Catalog # MFLX76000-08
Supplier: Avantor Fluid Handling
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Catalog # MFLX76000-08
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.
- No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
- Single-use pump heads are constructed with a disposable polypropylene chamber
- Reusable pump heads are constructed of durable 316L stainless steel
Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.
Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.
Specifications
- Description:Mutiple-Use Pump Chamber; 316L machined stainless steel multiuse Quattroflow pump systems
- Seq:04
- Cat. No.:MFLX76000-08
- Warranty:1 year
- Inlet connection:3⁄4" Tri-Clamp
- Wetted materials:316L SS (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
- Outlet connection:3⁄4" Tri-Clamp
Accessories
Product Information
- Catalog #Electrical plug typeMaximum Flow RateAvailabilityPrice
- Multi-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VACMulti-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VACMFLX74000-49No plug3 L/minLog in for availability1 * 1 ST - 13 250,00 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



