Order Entry
Belgium
Orders LinkContactUsLinkComponent
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems

Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems

Multiple-Use Pump Chamber; 316L machined for ABS and stainless steel multiuse Quattroflow pump systems
Catalog # MFLX76000-06
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Catalog # MFLX76000-06
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems

Multiple-Use Pump Chamber; 316L machined for ABS and stainless steel multiuse Quattroflow pump systems

Price
2 680,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.

  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Single-use pump heads are constructed with a disposable polypropylene chamber
  • Reusable pump heads are constructed of durable 316L stainless steel

Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.

Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.

Specifications
  • Description:
    Multiple-Use Pump Chamber; 316L machined for ABS and stainless steel multiuse Quattroflow pump systems
  • Seq:
    03
  • Cat. No.:
    MFLX76000-06
  • Warranty:
    1 year
  • Inlet connection:
    1⁄4" Tri-Clamp
  • Wetted materials:
    316L SS (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
  • Outlet connection:
    1⁄4" Tri-Clamp
Accessories

Product Information

  • Catalog #
    Electrical plug type
    Maximum Flow Rate
    Availability
    Price
  • Multi-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VAC
    Multi-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VAC
    MFLX74000-49
    No plug
    3 L/min
    Log in for availability
    1 * 1 ST - 13 250,00 €

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.