ContactUsLinkComponent
Masterflex® Fitting, 316 Stainless Steel, Tee, Hosebarb to Thread, 1/4" ID x 1/4" ID x 3/8" NPT(M)
Masterflex® Fitting, 316 Stainless Steel, Tee, Hosebarb to Thread, 1/4" ID x 1/4" ID x 3/8" NPT(M)
Catalog # MFLX31209-29
undefined
Masterflex® Fitting, 316 Stainless Steel, Tee, Hosebarb to Thread, 1/4" ID x 1/4" ID x 3/8" NPT(M)
Catalog # MFLX31209-29
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/4" ID x 1/4" ID x 3/8" NPT(M)
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
  • Barb size/tubing int.Ø (port 2):
    1/4"
  • Connection Type:
    Thread to Barb
  • Flow pattern:
    T
  • Description:
    Fitting, 316 Stainless Steel, Tee, Hosebarb to Thread, 1/4" ID x 1/4" ID x 3/8" NPT(M)
  • Seq:
    12
  • Environmentally Preferable:
  • Model:
    31209-29
  • Orifice dimensions:
    0,1875"
  • Warranty:
    1 year
  • Pk:
    1
  • Barb size/tubing int.Ø (port 3):
    1/4"
  • Fitting material:
    Stainless Steel 316
Frequently Bought Together