ContactUsLinkComponent
Masterflex® Fitting, Nylon, Straight, Threaded Reducer, 1/8" NPT(M) x 1/2" NPT (M); 6/PK
Masterflex® Fitting, Nylon, Straight, Threaded Reducer, 1/8" NPT(M) x 1/2" NPT (M); 6/PK
Catalog # MFLX30623-46
undefined
Masterflex® Fitting, Nylon, Straight, Threaded Reducer, 1/8" NPT(M) x 1/2" NPT (M); 6/PK
Catalog # MFLX30623-46
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/8" NPT(M) x 1/2" NPT (M)
  • Constructed of durable nylon
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.
  • Connection Type:
    Threaded Reducer
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ⅛"
  • Thread size NPT(M) (port 2):
    ½"
  • Description:
    Fitting, Nylon, Straight, Threaded Reducer, ⅛" NPT(M) x ½" NPT (M); 6/PK
  • Seq:
    13
  • Environmentally Preferable:
  • Model:
    30623-46
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Pk:
    6
  • Fitting material:
    Nylon
Frequently Bought Together