ContactUsLinkComponent
Masterflex® Footswitch for ON/OFF Pump Operation; 1/4" Phone Jack Plug Type
Masterflex® Footswitch for ON/OFF Pump Operation; 1/4" Phone Jack Plug Type
Catalog # MFLX07595-35
undefined
Masterflex® Footswitch for ON/OFF Pump Operation; 1/4" Phone Jack Plug Type
Catalog # MFLX07595-35
About this item
Make the most of your Masterflex® pump with connection cables, replacement parts, and other accessories.
  • Choose accessories and replacement parts suitable for your Masterflex® pump
  • Easily control your Masterflex® drive with compatible footswitch
Convenient accessories to make your job easier.
  • Description:
    Footswitch; 1/4" phone jack plug type, for on/off operation of drives.
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together