ContactUsLinkComponent
Masterflex® Touch Screen Syringe Pump Microliter Rack, for 4 Syringes
Masterflex® Touch Screen Syringe Pump Microliter Rack, for 4 Syringes
Catalog # MFLX74905-62
undefined
Masterflex® Touch Screen Syringe Pump Microliter Rack, for 4 Syringes
Catalog # MFLX74905-62
About this item
Make the most of your Masterflex® syringe pumps with these accessories.
  • Optimize performance of your Masterflex pumps
Parts and accessories for Masterflex syringe pumps.
  • Description:
    Syringe Pump Microliter Rack
  • Environmentally Preferable:
  • Model:
    74905-62
  • Syringe size accepted:
    0,5 µl - 10 ml
  • Warranty:
    1 year
  • Pk:
    1
  • No. of syringes:
    4
Frequently Bought Together