ContactUsLinkComponent
Masterflex® Syringe Pump Daisy Chain Cable for 74901-80 Pump
Masterflex® Syringe Pump Daisy Chain Cable for 74901-80 Pump
Catalog # MFLX74901-87
undefined
Masterflex® Syringe Pump Daisy Chain Cable for 74901-80 Pump
Catalog # MFLX74901-87
About this item
Daisy chain up to 100 pumps.
  • Connect multiple independent syringes to operate simultaneously
  • Easily connect pumps
  • Choose cable based on your current pump
  • For use with 74901-80 Dual Drive Syringe Pump
Optimize performance of your Masterflex® Dual-Drive Syringe Pump with these accessories.
  • Description:
    Pumps, Syringe Pumps Daisy Chain Cable for 74901-80 Dual Drive Syringe Pump
  • Seq:
    01
  • Environmentally Preferable:
  • Model:
    74901-87
  • Outlet connection:
    Daisy Chain Cable for 74901-80
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together