Order Entry
Belgium
Orders LinkContactUsLinkComponent
Accessories for Continuous Level Control, SymLine®

Accessories for Continuous Level Control, SymLine®

Built-in signal box table stand, brushed stainless steel, 2 mm wall thickness
Catalog # SCTV106698
Supplier:   S.C.A.T.
Accessories for Continuous Level Control, SymLine®
Catalog # SCTV106698
Supplier:  S.C.A.T.
Accessories for Continuous Level Control, SymLine®
Accessories for Continuous Level Control, SymLine®

Built-in signal box table stand, brushed stainless steel, 2 mm wall thickness

Price
61,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

LED's and acoustic alarm warn you in time before the fill level of your collection containers reaches the critical level. With mute / snooze function and interface for controlling external devices. For installation in your media line of the fume cupboard.

  • Built-in Signalbox2 for Safety Waste Cap and other electronical level controlers
  • Touchless alarm-resetting fields for two containers
  • Two switch inputs and two switch outputs for peripheral units
  • Incl. 230 V power supply cable with open end and clamp

Suitable e.g. for equipment from Wesemann, Waldner, Bense, Hemling, Köttermann, Kewaunee, Carlo Erba, Modus Complete, Delagrave, and many more.

Specifications
  • Description:
    Built-in signal box table stand, brushed stainless steel, 2 mm wall thickness
  • Seq:
    4
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.