ContactUsLinkComponent
Connection Box 100, Connection box for simplified installation and control of ATEX VARIO chemistry diaphragm pumps with vacuum controller (VACUU·SELECT or CVC 3000) and one vacuum sensor (both are available separately) 1 * 1 items
Catalog # VACU20635435
Supplier:  VACUUBRAND
Connection Box 100, Connection box for simplified installation and control of ATEX VARIO chemistry diaphragm pumps with vacuum controller (VACUU·SELECT or CVC 3000) and one vacuum sensor (both are available separately) 1 * 1 items
Catalog # VACU20635435
Supplier:  VACUUBRAND
Frequently Bought Together