Recombinant Human ASAM Protein (His Tag) 1 * 100 µG
Catalog # SINO10794-H08H-100
Supplier: Sino Biological
Recombinant Human ASAM Protein (His Tag) 1 * 100 µG
Catalog # SINO10794-H08H-100
Supplier: Sino Biological
Frequently Bought Together
J.T.Baker®, Plate 96
Supplier: Avantor
Storage plates, 384-well, MASTERBLOCK®
Supplier: Greiner Bio-One
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
Cryo.S™ tubes, 1 and 2 ml
Supplier: Greiner Bio-One
VWR®, Microcentrifuge Tubes and Screw Caps
Supplier: VWR Collection
Media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
Cryogenic tubes, CryoTubes™, Nunc™
Supplier: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cryoboxes, 133×133 mm
Supplier: TENAK
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
96-Well cell culture plates, Falcon®
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Whatman™ Cellulose Blotting Papers, Grade GB003, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)