ContactUsLinkComponent
Obtained from proprietary development, the precast gel SERVAGel TG PRiME 12 % features an extended shelf life and short electrophoresis times by using a standard Tris/glycine buffer system (2 gels, 35 min at 250 V const.). It can be operated under 1 * 10 items
  SERA43267.01
 :  Serva
Obtained from proprietary development, the precast gel SERVAGel TG PRiME 12 % features an extended shelf life and short electrophoresis times by using a standard Tris/glycine buffer system (2 gels, 35 min at 250 V const.). It can be operated under 1 * 10 items
  SERA43267.01
 :  Serva
Frequently Bought Together