ContactUsLinkComponent
Chemicals Valve Kit for Dosing Pumps 78167-00 to -10
Chemicals Valve Kit for Dosing Pumps 78167-00 to -10
Catalog # MFLX78167-63
1 year warranty
undefined
Chemicals Valve Kit for Dosing Pumps 78167-00 to -10
Catalog # MFLX78167-63
1 year warranty
About this item
Replacement parts keep your dosing pump running smoothly.
  • Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps
For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.
  • Description:
    Chemicals Valve Kit for Dosing Pumps 78167-00 to -10
  • Environmentally Preferable:
  • Wetted materials:
    FFKM
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together