Order Entry
Belgium
ContactUsLinkComponent
Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 1m, 1 Fitting
Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 1m, 1 Fitting
Catalog # MFLX78167-55
1 year warranty
undefined
Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 1m, 1 Fitting
Catalog # MFLX78167-55
1 year warranty

About this item

Replacement parts keep your dosing pump running smoothly.

  • Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps

For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.

  • Description:
    Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 1m, 1 Fitting
  • Environmentally Preferable:
  • Wetted materials:
    FEP
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together