ContactUsLinkComponent
Masterflex® luer fitting, male luer lock x 3/16 hose barb, Yellow, 25/pk"
Masterflex® luer fitting, male luer lock x 3/16 hose barb, Yellow, 25/pk"
Catalog # MFLX45520-25
undefined
Masterflex® luer fitting, male luer lock x 3/16 hose barb, Yellow, 25/pk"
Catalog # MFLX45520-25
About this item
Quick and easy assembly without tools.
  • Constructed of durable Polyolefin
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
Superior chemical resistance compared to polycarbonate. Sterilize by autoclave, gamma irradiation, or ethylene oxide. Meets USP Class VI. Animal-derived ingredient-free. Packs of 25.
  • Barb size/tubing int.Ø (port 1):
    3/16"
  • Connection Type:
    Male Luer to Barb
  • Flow pattern:
    Straight
  • Description:
    luer fitting, male luer lock×3/16 hose barb, Yellow, 25/pk"
  • Environmentally Preferable:
  • Connection type (port 1):
    Male luer lock
  • No. of ports:
    2
  • Pk:
    25
  • Fitting material:
    CrystalVu
Frequently Bought Together