ContactUsLinkComponent
Cadence Stainless steel stopcocks Female luer x female luer x male luer lock
Cadence Stainless steel stopcocks Female luer x female luer x male luer lock
Catalog # MFLX41607-02
undefined
Cadence Stainless steel stopcocks Female luer x female luer x male luer lock
Catalog # MFLX41607-02
About this item
Exceptional durability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured in an ISO 14644-1 Level 8 (formerly Class 100,000) clean room
  • Full material traceability on all items
Easily connect your tubing to your system with these durable luer stopcocks.
  • Description:
    Stainless steel stopcocks Female luer x female luer x male luer lock. 3 months warranty
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Connection type (port 1):
    Female luer
  • Connection type (port 2):
    Female luer
  • Connection type (port 3):
    Male luer lock
  • No. of ports:
    3
  • Pk:
    1
Frequently Bought Together