Order Entry
Belgium
ContactUsLinkComponent
Flowline LU10-2305 Ultrasonic Level Switch-PFA, Short, 3/4" NPT
Flowline LU10-2305 Ultrasonic Level Switch-PFA, Short, 3/4" NPT
Catalog # MFLX23002-16
IP65 washdown
undefined
Flowline LU10-2305 Ultrasonic Level Switch-PFA, Short, 3/4" NPT
Catalog # MFLX23002-16
IP65 washdown

About this item

Compatible with Smart Trak and Switch-Pak installation fittings.

  • CSA approved intrinsically safe for use in hazardous environments
  • 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity

The LU10 series ultrasonic level switch is an excellent choice for a broad range of liquids, including those with light coating or scaling-type characteristics. Liquid examples include xylene, chromic acid and light cooking oil.

  • Max. pressure:
    150 psi
  • Description:
    Ultrasonic Level Switch-PFA, Short, 3/4" NPT
  • Environmentally Preferable:
  • Wetted materials:
    PFA
  • Max. process temperature:
    80 °C
  • Min. process temperature:
    -40 °F
  • Relay output:
    SPST relay rated for 60 VA
  • Warranty:
    1 year
  • Pk:
    1
  • Power:
    12 - 36 VDC
Frequently Bought Together