ContactUsLinkComponent
Masterflex® Dispensing Tip with Luer Lock, Glass, Luer Lock by 3/16" and 1/4" ID
Masterflex® Dispensing Tip with Luer Lock, Glass, Luer Lock by 3/16" and 1/4" ID
Catalog # MFLX78226-60
undefined
Masterflex® Dispensing Tip with Luer Lock, Glass, Luer Lock by 3/16" and 1/4" ID
Catalog # MFLX78226-60
About this item
Make the most of your Masterflex® pump with connection cables, replacement parts, and other accessories.
  • Choose accessories and replacement parts suitable for your Masterflex® pump
  • Constructed of glass
  • Easily attach flexible tubing to the hose barb connection
Convenient accessories to make your job easier.
  • Description:
    Glass dispensing tip with Luer lock for 3/16" and 1/4" ID tubing.
  • Environmentally Preferable:
  • Pk:
    1
Frequently Bought Together