ContactUsLinkComponent
Masterflex® 6/10 Rack for Syringe Pump, Infusion and Withdrawal Design, Touchscreen
Masterflex® 6/10 Rack for Syringe Pump, Infusion and Withdrawal Design, Touchscreen
Catalog # MFLX74905-60
undefined
Masterflex® 6/10 Rack for Syringe Pump, Infusion and Withdrawal Design, Touchscreen
Catalog # MFLX74905-60
About this item
Make the most of your Masterflex® syringe pumps with these accessories.
  • Optimize performance of your Masterflex pumps
Parts and accessories for Masterflex syringe pumps.
  • Description:
    6/10 Rack for Syringe Pump, Infusion and Withdrawal design, Touchscreen
  • Environmentally Preferable:
  • Model:
    74905-60
  • Syringe size accepted:
    0,5 µl - 60 ml
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together