Micropump® Eagle Drive™ Pump Systems
Product Details & Documents
Complete pumps include head, motor, and integrated drive.
- Variable-speed electronic controller delivers smooth, pulseless flow
- Static seals reduce maintenance
- Small envelope-ideal for OEM use
Electromagnetically driven, precision-geared pumps provide smooth, accurate, pulseless flow. Torque improvements and compact size are achieved with NdFeB magnets. Control speed by using a 0 to 5 VDC input signal. Drives feature thermal and overload protection.
Choose a system with a suction shoe pump head to self compensate for wear, temperature, and pressure, and to maintain high volumetric efficiency. Cavity-style pump heads deliver higher lift capability and moderate pressure capability; an internal bypass is an integral safety feature of the cavity-style pump heads.
Delivery information: Includes 2-ft (0.6-m) power cord and screwdriver. Suction shoe service kits include: gears, suction shoe, shafts, scales, and springs. Cavity-style service kits include: gears, bushings, and seals.
Specifications
- Flow rate:0,91 ml/rev
- Maximum Flow Rate:4,55 L/min (1,2 gal./min)
- Description:Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0,91 ml/rev
- Cat. No.:MFLX74445-25
- Warranty:1 year
Accessories
Product Information
- Catalog #AvailabilityPrice
- Gear Pump Service Kit, for 74445-05
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



