ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Straight, Female Luer to Cap with Tether Loop Adapter; 25/PK
Masterflex® Fitting, Polypropylene, Straight, Female Luer to Cap with Tether Loop Adapter; 25/PK
Catalog # MFLX45508-64
CE certificate
undefined
Masterflex® Fitting, Polypropylene, Straight, Female Luer to Cap with Tether Loop Adapter; 25/PK
Catalog # MFLX45508-64
CE certificate
  • Model:
  • 45508-64
  • 30800-12
  • Connection type (port 1):
  • Female luer
  • Female luer
  • Connection type (port 2):
  • Cap with tether loop
  • Cap
  • No. of ports:
  • 1
  • 2
  • Warranty:
  • 3 Months
  • Currently Viewing
Specifications
About this item
Full material traceability to ensure process integrity.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Connection Type: Female Luer to Cap with Tether Loop
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
  • Connection Type:
    Female Luer Cap
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polypropylene, Straight, Female Luer to Cap with Tether Loop Adapter; 25/PK
  • Seq:
    02
  • Environmentally Preferable:
  • Model:
    45508-64
  • Connection type (port 1):
    Female luer
  • Connection type (port 2):
    Cap with tether loop
  • No. of ports:
    1
  • Pk:
    25
  • Fitting material:
    Polypropylene
Frequently Bought Together