ContactUsLinkComponent
Masterflex® Fitting, Nylon, Straight, Female Luer to Hose Barb Adapter, 1/8" ID; 25/PK
  MFLX45502-05
CE certificate
undefined
Masterflex® Fitting, Nylon, Straight, Female Luer to Hose Barb Adapter, 1/8" ID; 25/PK
  MFLX45502-05
CE certificate
  • Connection type (port 1):
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  •  
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance
  • Connection Type: Female Luer x 1/8" ID
  • Ideal for enhanced fluid path flow
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
  • Barb size/tubing int.Ø (port 1):
    1/8"
  • Connection Type:
    Female Luer to Barb
  • Flow pattern:
    Straight
  • Description:
    Fitting, Nylon, Straight, Female Luer to Hose Barb Adapter, 1/8" ID; 25/PK
  • Seq:
    06
  • Environmentally Preferable:
  • Connection type (port 1):
    Female luer
  • No. of ports:
    2
  • Warranty:
    3 Months
  • Pk:
    25
  • Fitting material:
    Nylon
Frequently Bought Together