ContactUsLinkComponent
Masterflex® Fitting, Polycarbonate, Straight, Female Luer to Barb Hose Adapter, 3/32" ID; 25/PK
Masterflex® Fitting, Polycarbonate, Straight, Female Luer to Barb Hose Adapter, 3/32" ID; 25/PK
Catalog # MFLX45501-02
CE certificate
Avantor exclusive
undefined
Masterflex® Fitting, Polycarbonate, Straight, Female Luer to Barb Hose Adapter, 3/32" ID; 25/PK
Catalog # MFLX45501-02
CE certificate
Avantor exclusive
About this item
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: Female Luer to Barb Hose Adapter, 3/32" ID
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
  • Barb size/tubing int.Ø (port 1):
    3/32"
  • Connection Type:
    Female Luer to Barb
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polycarbonate, Straight, Female Luer to Barb Hose Adapter, 3/32" ID; 25/PK
  • Seq:
    03
  • Environmentally Preferable:
  • Model:
    45501-02
  • Connection type (port 1):
    Female luer
  • No. of ports:
    2
  • Warranty:
    3 Months
  • Pk:
    25
  • Fitting material:
    Polycarbonate
Frequently Bought Together