ContactUsLinkComponent
Masterflex® Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 1/2" ID x 3/8" NPT(M)
Masterflex® Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 1/2" ID x 3/8" NPT(M)
Catalog # MFLX34030-80
CE certificate
undefined
Masterflex® Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 1/2" ID x 3/8" NPT(M)
Catalog # MFLX34030-80
CE certificate
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/2" ID x 3/8" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
  • Barb size/tubing int.Ø (port 2):
    1/2"
  • Connection Type:
    Thread to Barb
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    3/8"
  • Description:
    Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 1/2" ID x 3/8" NPT(M)
  • Seq:
    08
  • Environmentally Preferable:
  • No. of ports:
    2
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Warranty:
    2 years
  • Pk:
    1
  • Fitting material:
    PTFE
Frequently Bought Together