ContactUsLinkComponent
Masterflex® Fitting, Black PVDF, One-Way Stopcock, Male Luer Lock to Male Luer Lock; 10/PK
Masterflex® Fitting, Black PVDF, One-Way Stopcock, Male Luer Lock to Male Luer Lock; 10/PK
Catalog # MFLX31200-90
undefined
Masterflex® Fitting, Black PVDF, One-Way Stopcock, Male Luer Lock to Male Luer Lock; 10/PK
Catalog # MFLX31200-90
About this item
Build leakproof fluid handling systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
These one-way stopcocks feature an all-black PVDF construction.
  • Flow pattern:
    One-Way
  • Description:
    Luer stopcock, 1-way, male luer lock x male luer lock, black PVDF, pack of 10
  • Seq:
    02
  • Environmentally Preferable:
  • Model:
    31200-90
  • Wetted materials:
    Black PVDF
  • Pk:
    10
  • Inlet port:
    Male Luer Lock
  • Outlet port:
    Male Luer Lock
Frequently Bought Together