ContactUsLinkComponent
Masterflex® Torque Tee Sanitary Clamp Securing Device, Swivel Joint, ¹/₄"; 1/EA
Masterflex® Torque Tee Sanitary Clamp Securing Device, Swivel Joint, ¹/₄"; 1/EA
Catalog # MFLX23508-05
1 year warranty
undefined
Masterflex® Torque Tee Sanitary Clamp Securing Device, Swivel Joint, ¹/₄"; 1/EA
Catalog # MFLX23508-05
1 year warranty
  • Drive:
  • 0,25"
  • 40"/lb
  • 0,25"
  • 0,25"
  • 0,875" (50" lb.)
  • 0,375"
  • 0,25"
  • 0,25" (50" lb.)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • Currently Viewing
Specifications
About this item
Ensure the security of sanitary connections with preset torque output.
  • Works with any sanitary clamp wing nut and Rubberfab's Torque-Rite® nut
  • Easily reach into tight or awkward spaces
  • Choose the appropriate torque output per the clamp manufacturer's specifications
  • Audible click alerts you when the torque has been reached
  • Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
  • 'T' shape creates effective leverage for fast, easy one-handed tightening
The Rubberfab Torque Tee and universal socket are designed to safely and securely install and remove sanitary clamps. Available with a family of components, the Torque Tee handle creates effective tightening leverage, even in hard-to-reach spaces. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle. The universal socket is required when using the Torque Tee.
  • Description:
    Torque Tee Sanitary Clamp Securing Device, Swivel Joint, ¹/₄"; 1/EA
  • Seq:
    03
  • Environmentally Preferable:
  • Pk:
    1
  • Fitting material:
    Stainless Steel
  • Drive:
    0,25"
  • Warranty:
    1 year
Frequently Bought Together