Order Entry
Belgium
ContactUsLinkComponent
PendoTech PMAT-CB-MF25 Single-Use Pressure Mat Interface Cable; 6 ft (1.8 m)
PendoTech PMAT-CB-MF25 Single-Use Pressure Mat Interface Cable; 6 ft (1.8 m)
Catalog # MFLX19406-87
CE Compliant
undefined
PendoTech PMAT-CB-MF25 Single-Use Pressure Mat Interface Cable; 6 ft (1.8 m)
Catalog # MFLX19406-87
CE Compliant

About this item

Optimize performance of your PendoTECH® Pressure Sensor.

  • Choose from a wide range of replacement parts and accessories

Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.

Use this cable to connect to your PendoTECH PressureMAT™ relay output to any Masterflex® Drive with a DB25 remote I/O connector. Allows for PMAT to utilize Start/Stop control of pump.

  • Length:
    182,88 cm (72")
  • Description:
    Interface cable 6 ft (1,8 m)
  • Environmentally Preferable:
  • Warranty:
    2 years
  • Pk:
    1
Frequently Bought Together