PendoTECH® Single-Use Pressure Sensor Accessories
RS232 Sensor Cable with 4-wire leads, 12 ft (3,7 m)
Catalog # MFLX19406-82
Supplier: Avantor Fluid Handling
PendoTECH® Single-Use Pressure Sensor Accessories
Catalog # MFLX19406-82
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:RS232 Sensor Cable with 4-wire leads, 12 ft (3,7 m)
- Cat. No.:MFLX19406-82
- Warranty:1 year
- Length:365,76 cm (144")



