PendoTECH® Single-Use Pressure Sensor Accessories
Sensor Cable with 4-wire leads, for 3rd party monitors 12ft (3,7 m)
Catalog # MFLX19406-80
Supplier: Avantor Fluid Handling
PendoTECH® Single-Use Pressure Sensor Accessories
Catalog # MFLX19406-80
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Sensor Cable with 4-wire leads, for 3rd party monitors 12ft (3,7 m)
- Cat. No.:MFLX19406-80
- Garantie:1 year
- Longueur:365,76 cm (144")
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



