ContactUsLinkComponent
Masterflex® L/S® Gear Service Kits for 1 to 100 rpm Drives
Masterflex® L/S® Gear Service Kits for 1 to 100 rpm Drives
Catalog # MFLX07553-08
undefined
Masterflex® L/S® Gear Service Kits for 1 to 100 rpm Drives
Catalog # MFLX07553-08
About this item
Make the most of your Masterflex drives replacement parts and service kits.
  • Easy to install, these service kits avoid costly downtime of your gear pump head
  • Suitable for use with Masterflex® L/S® 1 to 100 rpm drives
Extend the life of your Masterflex pump with these replacement parts.
Delivery: Includes two gear assemblies (large and small), two gear case cover gaskets, and one vial of high temperature silicone grease.
  • Description:
    Service kits for L/S 1-100 rpm drives.
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Compatible with:
    07520-10, -30, -35, -50, -55, 07521-10, -30, -35, -50, -55, 07522-00, -05, 07523-10, -12, -30, -35, 07525-10, -12, -30, -32, 07527-10, -12, 07531-10,
  • Pk:
    1
Frequently Bought Together