ContactUsLinkComponent
Masterflex® DB25 Male Connector; for Creating Your Own Cable
Masterflex® DB25 Male Connector; for Creating Your Own Cable
Catalog # MFLX07523-94
undefined
Masterflex® DB25 Male Connector; for Creating Your Own Cable
Catalog # MFLX07523-94
About this item
Create your own cable for optimization of your Masterflex® drive.
  • Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).
  • Description:
    DB25 Male connector; for creating your own cable
  • Environmentally Preferable:
  • Model:
    07523-94
  • Control type:
    Analog Variable Speed
  • Warranty:
    2 years
  • Pk:
    1
Frequently Bought Together