ContactUsLinkComponent
Masterflex® L/S® Easy-Load® Pump Head, Replacement Rotor and Bearing Assembly, White PPS Housing, CRS Rotor, for 07516-00
Masterflex® L/S® Easy-Load® Pump Head, Replacement Rotor and Bearing Assembly, White PPS Housing, CRS Rotor, for 07516-00
Catalog # MFLX07516-08
undefined
Masterflex® L/S® Easy-Load® Pump Head, Replacement Rotor and Bearing Assembly, White PPS Housing, CRS Rotor, for 07516-00
Catalog # MFLX07516-08
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® and I/P® pump head
  • Suitable for Easy-Load® Pump Heads 07516-00 and 07518-00
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
  • Pump series:
    L/S
  • Description:
    Easy-Load® Pump Head, Replacement Rotor and Bearing Assembly, White PPS Housing, CRS Rotor; 1/Ea
  • Seq:
    38
  • Environmentally Preferable:
  • Model:
    07516-08
  • Open-head sensor:
    No
  • Warranty:
    2 years
  • Pk:
    1
  • Housing material:
    PPS
  • Rotor material:
    Cold-rolled steel (CRS)
Frequently Bought Together