ContactUsLinkComponent
Masterflex® L/S® Easy-Load® Pump Head Replacement Rotor/Bearing Assembly for 07514-10 and -30
Masterflex® L/S® Easy-Load® Pump Head Replacement Rotor/Bearing Assembly for 07514-10 and -30
Catalog # MFLX07514-18
1 year warranty
undefined
Masterflex® L/S® Easy-Load® Pump Head Replacement Rotor/Bearing Assembly for 07514-10 and -30
Catalog # MFLX07514-18
1 year warranty
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
  • Pump series:
    L/S
  • Description:
    L/S® Easy-Load® Pump Head Replacement Rotor/Bearing Assembly for 07514-10 and -30
  • Seq:
    36
  • Environmentally Preferable:
  • Model:
    07514-18
  • Open-head sensor:
    No
  • Tubing sizes accepted:
    L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18
  • Warranty:
    1 year
  • Pk:
    1
  • Rollers:
    4
  • Rotor material:
    Stainless Steel
Frequently Bought Together