ContactUsLinkComponent
Masterflex® I/P® Replacement Rotor Assembly, Long-Shaft, CRS
Masterflex® I/P® Replacement Rotor Assembly, Long-Shaft, CRS
Catalog # MFLX07019-80
undefined
Masterflex® I/P® Replacement Rotor Assembly, Long-Shaft, CRS
Catalog # MFLX07019-80
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
  • Pump series:
    I/P
  • Description:
    Replacement Rotor Assembly, Long-Shaft, CRS
  • Seq:
    25
  • Environmentally Preferable:
  • Model:
    07019-80
  • Open-head sensor:
    No
  • Warranty:
    2 years
  • Pk:
    1
  • Rotor material:
    Cold-rolled steel (CRS)
Frequently Bought Together