ContactUsLinkComponent
Masterflex® L/S® Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, CRS Rotor, L/S® 14
Masterflex® L/S® Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, CRS Rotor, L/S® 14
Catalog # MFLX07014-81
undefined
Masterflex® L/S® Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, CRS Rotor, L/S® 14
Catalog # MFLX07014-81
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® and I/P® pump head
  • Suitable for use with L/S® Standard Pump Heads 07014-00, -20, -70, and -72
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
  • Pump series:
    L/S
  • Description:
    Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, CRS Rotor, L/S® 14 Tubing
  • Seq:
    06
  • Environmentally Preferable:
  • Model:
    07014-81
  • Open-head sensor:
    No
  • Tubing sizes accepted:
    L/S 14
  • Warranty:
    2 years
  • Pk:
    1
  • Housing material:
    PC
  • Rotor material:
    Cold-rolled steel (CRS)
Frequently Bought Together