Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems

Disposable Pump Chamber; PP machined for stainless steel single-use Quattroflow pump systems
  MFLX76000-02
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
  MFLX76000-02
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems

Disposable Pump Chamber; PP machined for stainless steel single-use Quattroflow pump systems

 
2 310,00 €
   
1 * 1 ST
 

Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.

  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Single-use pump heads are constructed with a disposable polypropylene chamber
  • Reusable pump heads are constructed of durable 316L stainless steel

Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.

Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.

 
  • Description:
    Disposable Pump Chamber; PP machined for stainless steel single-use Quattroflow pump systems
  • Seq:
    02
  • Cat. No.:
    MFLX76000-02
  • Warranty:
    Shelf life
  • Inlet connection:
    3⁄4" Tri-Clamp
  • Wetted materials:
    PP (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
  • Outlet connection:
    3⁄4" Tri-Clamp
Accessories

 

  •  
    Description
    Electrical plug type
    Maximum Flow Rate
     
     
  • MFLX74000-49
    Multi-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VAC
    No plug
    3 L/min
       
    1 * 1 ST - 13 250,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.